siRNA Details
| Gene Symbol: LOC256880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125923.1 | 4 | 457 | |||
| GenBank mRNA | DB462285.1 | - | 4 | 428 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC256880| Gene Symbol: LOC256880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125923.1 | 4 | 457 | |||
| GenBank mRNA | DB462285.1 | - | 4 | 428 | ||