siRNA Details
| Gene Symbol: LINC01366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC093717.1 | - | 1 | 252 | ||
| BC111994.1 | - | 1 | 252 | |||
| BQ053612.1 | - | 3 | 742 | |||
| CD518314.1 | - | 3 | 724 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01366| Gene Symbol: LINC01366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC093717.1 | - | 1 | 252 | ||
| BC111994.1 | - | 1 | 252 | |||
| BQ053612.1 | - | 3 | 742 | |||
| CD518314.1 | - | 3 | 724 | |||