siRNA Details
| Gene Symbol: NCR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145466.1 | NP_001138938.1 | 1 | 278 | ||
| NM_001145467.1 | NP_001138939.1 | 1 | 278 | |||
| NM_147130.2 | NP_667341.1 | 1 | 278 | |||
| XM_006715049.3 | XP_006715112.1 | 2 | 224 | |||
| XM_011514459.2 | XP_011512761.1 | 1 | 317 | |||
| GenBank mRNA | AB055881.1 | - | 1 | 14 | ||
| AF031136.1 | - | 1 | 278 | |||
| AF031137.1 | - | 1 | 278 | |||
| AF031138.1 | - | 1 | 278 | |||
| AJ223153.1 | - | 1 | 77 | |||
| BC018752.1 | - | 1 | 242 | 4849421 | ||
| BC052582.1 | - | 1 | 222 | 5934651 | ||

NCR3
Genomic Map