siRNA Details
| Gene Symbol: NCR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145466.1 | NP_001138938.1 | 2 | 426 | ||
| NM_001145467.1 | NP_001138939.1 | 2 | 426 | |||
| NM_147130.2 | NP_667341.1 | 2 | 426 | |||
| XM_006715049.3 | XP_006715112.1 | 3 | 372 | |||
| XM_011514459.2 | XP_011512761.1 | 2 | 465 | |||
| GenBank mRNA | AB055881.1 | - | 2 | 162 | ||
| AF031136.1 | - | 2 | 426 | |||
| AF031137.1 | - | 2 | 426 | |||
| AF031138.1 | - | 2 | 426 | |||
| AJ223153.1 | - | 2 | 225 | |||
| BC018752.1 | - | 2 | 390 | 4849421 | ||
| BC052582.1 | - | 2 | 370 | 5934651 | ||
| BG744854.1 | - | 3 | 681 | |||

NCR3
Genomic Map