siRNA Details
| Gene Symbol: SSX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173358.2 | NP_775494.1 | 3, 4 | 335 | ||
| XM_017029430.1 | XP_016884919.1 | 2, 3 | 176 | |||
| XM_017029431.1 | XP_016884920.1 | 2, 3 | 176 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SSX7| Gene Symbol: SSX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173358.2 | NP_775494.1 | 3, 4 | 335 | ||
| XM_017029430.1 | XP_016884919.1 | 2, 3 | 176 | |||
| XM_017029431.1 | XP_016884920.1 | 2, 3 | 176 | |||