siRNA Details
| Gene Symbol: FLJ37201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026835.1 | 2 | 919 | |||
| GenBank mRNA | AK094520.1 | - | 1 | 74 | ||
| BC043217.2 | - | 2 | 919 | |||
| BC104181.2 | - | 1 | 73 | |||
| BC104182.2 | - | 1 | 73 | |||
| BC112983.1 | - | 1 | 73 | |||
| BC112984.1 | - | 1 | 73 | |||

FLJ37201
Genomic Map