siRNA Details
Gene Symbol: LOC107987205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017021847.1 | XP_016877336.1 | 1 | 1804 |
Gene Symbol: BMS1P17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DA741885.1 | - | 1 | 72 |
Gene Symbol: BMS1P22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_133911.1 | 1 | 72 |
Gene Symbol: DUXAP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC066547.1 | - | 1 | 1146 | 5481815 |
Gene Symbol: LINC01296 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096951.1 | - | 1 | 1670 |
Gene Symbol: LOC101929839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546155.1 | XP_011544457.1 | 1 | 1804 |
Gene Symbol: LOC283028 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136644.1 | 1 | 631 | |||
GenBank mRNA | AK092783.1 | - | 1 | 631 |
