siRNA Details
| Gene Symbol: ZMIZ1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015429.1 | 4 | 316 | |||
| NR_024431.2 | 6 | 608 | ||||
| GenBank mRNA | AK098249.1 | - | 6 | 1053 | ||
| BC102000.1 | - | 3 | 159 | |||
| BC102001.1 | - | 3 | 159 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZMIZ1-AS1| Gene Symbol: ZMIZ1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015429.1 | 4 | 316 | |||
| NR_024431.2 | 6 | 608 | ||||
| GenBank mRNA | AK098249.1 | - | 6 | 1053 | ||
| BC102000.1 | - | 3 | 159 | |||
| BC102001.1 | - | 3 | 159 | |||