siRNA Details
Gene Symbol: ZMIZ1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024429.1 | 4 | 416 | |||
NR_024431.2 | 5 | 549 | ||||
GenBank mRNA | AK098249.1 | - | 5 | 994 | ||
BC105703.1 | - | 3 | 259 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZMIZ1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024429.1 | 4 | 416 | |||
NR_024431.2 | 5 | 549 | ||||
GenBank mRNA | AK098249.1 | - | 5 | 994 | ||
BC105703.1 | - | 3 | 259 |