siRNA Details
| Gene Symbol: CDNF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029954.2 | NP_001025125.2 | 4 | 449 | ||
| XM_011519488.2 | XP_011517790.1 | 4 | 565 | |||
| GenBank mRNA | BC037872.1 | - | 6 | 614 | ||
| BC133042.1 | - | 4 | 436 | 40147064 | ||
| BC133044.1 | - | 4 | 436 | 40147066 | ||
| BC144619.1 | - | 4 | 421 | |||
| BG213461.1 | - | 1 | 29 | |||
| CN311321.1 | - | 4 | 425 | |||

CDNF
Genomic Map