siRNA Details
| Gene Symbol: CSNK2A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256686.1 | NP_001243615.1 | 1 | 14 | ||
| Gene Symbol: GALNT18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011520071.2 | XP_011518373.1 | 7 | 2709 | ||

CSNK2A3
Genomic Map