siRNA Details
| Gene Symbol: LOC283140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126004.1 | 4 | 1167 | |||
| GenBank mRNA | AK095275.1 | - | 4 | 1171 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC283140| Gene Symbol: LOC283140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126004.1 | 4 | 1167 | |||
| GenBank mRNA | AK095275.1 | - | 4 | 1171 | ||