siRNA Details
Gene Symbol: LOC283140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126004.1 | 4 | 1202 | |||
GenBank mRNA | AK095275.1 | - | 4 | 1206 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC283140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126004.1 | 4 | 1202 | |||
GenBank mRNA | AK095275.1 | - | 4 | 1206 |