siRNA Details
| Gene Symbol: KIRREL3-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040078.1 | 1 | 768 | |||
| GenBank mRNA | AK097543.1 | - | 1 | 768 | ||
| BC113593.1 | - | 1 | 275 | 8322645 | ||
| BC113597.1 | - | 1 | 275 | 8322649 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIRREL3-AS3| Gene Symbol: KIRREL3-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040078.1 | 1 | 768 | |||
| GenBank mRNA | AK097543.1 | - | 1 | 768 | ||
| BC113593.1 | - | 1 | 275 | 8322645 | ||
| BC113597.1 | - | 1 | 275 | 8322649 | ||