siRNA Details
| Gene Symbol: LOC283332 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026948.1 | 3 | 189 | |||
| GenBank mRNA | AK093215.1 | - | 3 | 183 | ||
| BC121095.1 | - | 1 | 254 | |||
| BC121096.1 | - | 2 | 136 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC283332| Gene Symbol: LOC283332 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026948.1 | 3 | 189 | |||
| GenBank mRNA | AK093215.1 | - | 3 | 183 | ||
| BC121095.1 | - | 1 | 254 | |||
| BC121096.1 | - | 2 | 136 | |||