siRNA Details
| Gene Symbol: LINC01465 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121682.1 | 1 | 453 | |||
| GenBank mRNA | AK098456.1 | - | 1 | 454 | ||
| BC038738.1 | - | 1 | 156 | |||
| BC136288.1 | - | 1 | 447 | 9020275 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01465| Gene Symbol: LINC01465 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121682.1 | 1 | 453 | |||
| GenBank mRNA | AK098456.1 | - | 1 | 454 | ||
| BC038738.1 | - | 1 | 156 | |||
| BC136288.1 | - | 1 | 447 | 9020275 | ||