siRNA Details
| Gene Symbol: FGF14-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036486.1 | 1, 2 | 172 | |||
| GenBank mRNA | AK125748.1 | - | 1, 2 | 172 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FGF14-IT1| Gene Symbol: FGF14-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036486.1 | 1, 2 | 172 | |||
| GenBank mRNA | AK125748.1 | - | 1, 2 | 172 | ||