siRNA Details
| Gene Symbol: GPR137C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099652.1 | NP_001093122.1 | 1 | 420 | ||
| GenBank mRNA | AK123953.1 | - | 1 | 316 | ||
| AL834372.1 | - | 1 | 280 | |||
| AW470753.1 | - | 1 | 359 | |||
| AY255590.1 | - | 1 | 327 | |||

GPR137C
Genomic Map