siRNA Details
| Gene Symbol: C14orf80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134875.1 | NP_001128347.1 | 8 | 1272 | ||
| NM_001134876.1 | NP_001128348.1 | 7 | 1173 | |||
| NM_001134877.1 | NP_001128349.1 | 8 | 1100 | |||
| NM_001198983.1 | NP_001185912.1 | 8 | 1182 | |||
| GenBank mRNA | AK024506.1 | - | 2 | 4014 | ||
| BC016028.1 | - | 7 | 940 | 3907551 | ||
| BX161477.1 | - | 8 | 1182 | |||
| BX248038.1 | - | 8 | 1100 | |||
| BX248074.1 | - | 9 | 1400 | |||
| BX248283.1 | - | 7 | 1131 | |||

C14orf80
Genomic Map