siRNA Details
Gene Symbol: LOC283710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243538.1 | NP_001230467.1 | 4 | 947 | ||
XM_011521465.2 | XP_011519767.1 | 5 | 804 | |||
GenBank mRNA | AA749020.1 | - | 1 | 90 |
