siRNA Details
| Gene Symbol: CASC22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135281.1 | 2, 3 | 101 | |||
| GenBank mRNA | AK093211.1 | - | 2, 3 | 1697 | ||
| AW772079.1 | - | 1, 2 | 281 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CASC22| Gene Symbol: CASC22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135281.1 | 2, 3 | 101 | |||
| GenBank mRNA | AK093211.1 | - | 2, 3 | 1697 | ||
| AW772079.1 | - | 1, 2 | 281 | |||