siRNA Details
| Gene Symbol: ERVK13-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040023.1 | 3 | 4651 | |||
| GenBank mRNA | AK000400.1 | - | 1 | 321 | ||
| AK091794.1 | - | 3 | 2443 | |||
| AK095440.1 | - | 1 | 2248 | |||
| AK097909.1 | - | 1 | 706 | |||
| AK131509.1 | - | 1 | 3277 | |||
| AK292678.1 | - | 1 | 1224 | |||
| AX747190.1 | - | 3 | 2443 | |||
| BC043252.1 | - | 3 | 4653 | |||

ERVK13-1
Genomic Map