siRNA Details
| Gene Symbol: LINC00921 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033904.1 | 1 | 797 | |||
| GenBank mRNA | AK096958.1 | - | 1 | 763 | ||
| BC104941.1 | - | 1 | 822 | |||
| BC104943.1 | - | 1 | 822 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00921| Gene Symbol: LINC00921 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033904.1 | 1 | 797 | |||
| GenBank mRNA | AK096958.1 | - | 1 | 763 | ||
| BC104941.1 | - | 1 | 822 | |||
| BC104943.1 | - | 1 | 822 | |||