siRNA Details
| Gene Symbol: LINC01566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027079.2 | 4, 5 | 803 | |||
| GenBank mRNA | BC045579.1 | - | 4, 5 | 803 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01566| Gene Symbol: LINC01566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027079.2 | 4, 5 | 803 | |||
| GenBank mRNA | BC045579.1 | - | 4, 5 | 803 | ||