siRNA Details
| Gene Symbol: TMEM235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204210.1 | NP_001191139.1 | 4 | 992 | ||
| NM_001204211.1 | NP_001191140.1 | 3 | 911 | |||
| XM_011524620.2 | XP_011522922.1 | 3 | 368 | |||
| XM_011524624.2 | XP_011522926.1 | 3 | 368 | |||
| XM_017024459.1 | XP_016879948.1 | 3 | 368 | |||
| XM_017024460.1 | XP_016879949.1 | 3 | 368 | |||
| XM_017024461.1 | XP_016879950.1 | 3 | 368 | |||
| GenBank mRNA | BI757060.1 | - | 3 | 330 | ||
| BI757981.1 | - | 3 | 262 | |||

TMEM235
Genomic Map