siRNA Details
| Gene Symbol: C17orf105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136483.1 | NP_001129955.1 | 4, 5 | 456 | ||
| XM_011524633.1 | XP_011522935.1 | 5, 6 | 1695 | |||
| GenBank mRNA | AK125817.1 | - | 4, 5 | 431 | ||
| BC054872.1 | - | 4, 5 | 460 | |||
| BC070324.1 | - | 4, 5 | 456 | |||
| BC146971.1 | - | 4, 5 | 455 | 9056899 | ||

C17orf105
Genomic Map