siRNA Details
| Gene Symbol: MEIOC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145080.2 | NP_001138552.2 | 5 | 1213 | ||
| XM_005257236.3 | XP_005257293.1 | 5 | 1190 | |||
| GenBank mRNA | AK093167.1 | - | 2 | 549 | ||
| AK302001.1 | - | 3 | 654 | |||
| BC035159.1 | - | 2 | 905 | 5265358 | ||
| BC132822.1 | - | 1 | 534 | 40146844 | ||
| BC136642.1 | - | 1 | 534 | 9020632 | ||

MEIOC
Genomic Map