siRNA Details
Gene Symbol: LINC00907 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046454.1 | 6 | 685 | |||
NR_046455.1 | 5 | 522 | ||||
GenBank mRNA | AB593073.1 | - | 5 | 522 | ||
BC040725.2 | - | 6 | 664 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00907 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046454.1 | 6 | 685 | |||
NR_046455.1 | 5 | 522 | ||||
GenBank mRNA | AB593073.1 | - | 5 | 522 | ||
BC040725.2 | - | 6 | 664 |