siRNA Details
| Gene Symbol: C19orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198476.3 | NP_940878.3 | 4 | 726 | ||
| XM_005258776.4 | XP_005258833.1 | 4 | 1306 | |||
| XM_006723152.2 | XP_006723215.1 | 4 | 1306 | |||
| XM_006723153.2 | XP_006723216.1 | 4 | 1306 | |||
| XM_011526774.2 | XP_011525076.1 | 4 | 1306 | |||
| XM_011526775.1 | XP_011525077.1 | 4 | 1306 | |||
| XM_011526776.2 | XP_011525078.1 | 4 | 1306 | |||
| XM_011526777.2 | XP_011525079.1 | 4 | 1306 | |||
| XM_011526778.2 | XP_011525080.1 | 4 | 1306 | |||
| XM_011526779.2 | XP_011525081.1 | 4 | 1306 | |||
| XM_011526782.2 | XP_011525084.1 | 4 | 1306 | |||
| XM_011526784.2 | XP_011525086.1 | 5 | 629 | |||
| XM_011526786.2 | XP_011525088.1 | 4 | 1306 | |||
| XM_017026620.1 | XP_016882109.1 | 4 | 1306 | |||
| XM_017026621.1 | XP_016882110.1 | 4 | 1306 | |||
| XM_017026622.1 | XP_016882111.1 | 4 | 1306 | |||
| XM_017026624.1 | XP_016882113.1 | 5 | 629 | |||
| XM_017026625.1 | XP_016882114.1 | 4 | 1306 | |||
| XM_017026626.1 | XP_016882115.1 | 5 | 629 | |||
| GenBank mRNA | AK123126.1 | - | 5 | 629 | ||
| BI521309.1 | - | 5 | 558 | |||
| CN281611.1 | - | 4 | 389 | |||

C19orf54
Genomic Map