siRNA Details
| Gene Symbol: PHLDB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198850.3 | NP_942147.3 | 6 | 947 | ||
| XM_005259169.3 | XP_005259226.1 | 6 | 1030 | |||
| XM_005259170.3 | XP_005259227.1 | 6 | 1030 | |||
| XM_005259171.3 | XP_005259228.1 | 6 | 1030 | |||
| XM_011527236.2 | XP_011525538.1 | 6 | 1134 | |||
| GenBank mRNA | AK097512.1 | - | 6 | 757 | ||
| BC070094.1 | - | 6 | 1049 | 4374205 | ||
| BU858992.1 | - | 4 | 471 | |||

PHLDB3
Genomic Map