siRNA Details
| Gene Symbol: EMC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175063.5 | NP_778233.4 | 6 | 730 | ||
| NM_206538.3 | NP_996261.1 | 6 | 730 | |||
| XM_006723163.3 | XP_006723226.1 | 6 | 724 | |||
| XM_011526818.2 | XP_011525120.1 | 6 | 724 | |||
| GenBank mRNA | AL512761.1 | - | 2 | 246 | ||
| AY194293.2 | - | 10 | 1688 | |||
| AY358710.1 | - | 6 | 690 | |||
| AY761095.1 | - | 6 | 708 | |||
| AY761097.1 | - | 6 | 708 | |||
| BC032948.2 | - | 6 | 671 | 4794992 | ||
| BC035001.2 | - | 6 | 669 | 4796476 | ||
| BC062607.1 | - | 6 | 872 | |||
| CN423491.1 | - | 3 | 355 | |||
| HY149334.1 | - | 3 | 364 | |||

EMC10
Genomic Map