siRNA Details
Gene Symbol: ZNF528-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK022990.1 | - | 1 | 1320 | ||
AK097590.1 | - | 5 | 836 | |||
BX537909.1 | - | 4 | 4775 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF528-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK022990.1 | - | 1 | 1320 | ||
AK097590.1 | - | 5 | 836 | |||
BX537909.1 | - | 4 | 4775 |