siRNA Details
| Gene Symbol: ZNF561-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122038.1 | 3 | 281 | |||
| GenBank mRNA | AK097685.1 | - | 3 | 246 | ||
| BC143540.1 | - | 2 | 154 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF561-AS1| Gene Symbol: ZNF561-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122038.1 | 3 | 281 | |||
| GenBank mRNA | AK097685.1 | - | 3 | 246 | ||
| BC143540.1 | - | 2 | 154 | |||