siRNA Details
| Gene Symbol: OR10H5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004466.1 | NP_001004466.1 | 1 | 339 | ||
| GenBank mRNA | BC136869.1 | - | 1 | 437 | 9020860 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR10H5| Gene Symbol: OR10H5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004466.1 | NP_001004466.1 | 1 | 339 | ||
| GenBank mRNA | BC136869.1 | - | 1 | 437 | 9020860 | |
Set Membership: |
|