siRNA Details
| Gene Symbol: LOC284581 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046098.1 | 1 | 406 | |||
| GenBank mRNA | AK094426.1 | - | 1 | 891 | ||
| AK096315.1 | - | 1 | 406 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC284581| Gene Symbol: LOC284581 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046098.1 | 1 | 406 | |||
| GenBank mRNA | AK094426.1 | - | 1 | 891 | ||
| AK096315.1 | - | 1 | 406 | |||