siRNA Details
| Gene Symbol: LINC01128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047519.1 | 3, 4 | 439 | |||
| GenBank mRNA | AK026873.1 | - | 3, 4 | 331 | ||
| BC102012.1 | - | 3, 4 | 314 | |||
| BC102023.1 | - | 3, 4 | 314 | |||
| BM459718.1 | - | 3, 4 | 312 | |||
| DR005509.1 | - | 3, 4 | 328 | |||
| DR005973.1 | - | 3, 4 | 351 | |||

LINC01128
Genomic Map