siRNA Details
Gene Symbol: LOC100130417 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026874.2 | 3 | 526 | |||
NR_122045.1 | 3 | 466 | ||||
GenBank mRNA | AK096928.1 | - | 1 | 2918 | ||
BC025320.1 | - | 1 | 572 | |||
BC144023.1 | - | 3 | 271 | |||
BC144024.1 | - | 3 | 211 |
