siRNA Details
| Gene Symbol: FAM102B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010883.2 | NP_001010883.2 | 4 | 432 | ||
| GenBank mRNA | BC127898.1 | - | 4 | 432 | 40132918 | |
| BC140863.1 | - | 4 | 288 | 9021731 | ||
| BC140865.1 | - | 4 | 288 | 9021733 | ||
| BC150530.1 | - | 4 | 432 | 40132919 | ||

FAM102B
Genomic Map