siRNA Details
| Gene Symbol: SMG7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040063.1 | 1 | 183 | |||
| GenBank mRNA | AI793165.1 | - | 1 | 200 | ||
| AL050020.1 | - | 1 | 470 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMG7-AS1| Gene Symbol: SMG7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040063.1 | 1 | 183 | |||
| GenBank mRNA | AI793165.1 | - | 1 | 200 | ||
| AL050020.1 | - | 1 | 470 | |||