siRNA Details
| Gene Symbol: LINC01270 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034124.1 | 2 | 315 | |||
| GenBank mRNA | AK090605.1 | - | 2 | 315 | ||
| BC093937.1 | - | 2 | 305 | 7939782 | ||
| BC093939.1 | - | 2 | 305 | 7939784 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01270| Gene Symbol: LINC01270 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034124.1 | 2 | 315 | |||
| GenBank mRNA | AK090605.1 | - | 2 | 315 | ||
| BC093937.1 | - | 2 | 305 | 7939782 | ||
| BC093939.1 | - | 2 | 305 | 7939784 | ||