siRNA Details
| Gene Symbol: LOC284825 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126010.1 | 4 | 812 | |||
| GenBank mRNA | AK093119.1 | - | 4 | 812 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC284825| Gene Symbol: LOC284825 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126010.1 | 4 | 812 | |||
| GenBank mRNA | AK093119.1 | - | 4 | 812 | ||