siRNA Details
| Gene Symbol: LOC284898 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318329.1 | NP_001305258.1 | 1 | 264 | ||
| GenBank mRNA | AK055980.1 | - | 1 | 44 | ||
| BC036876.1 | - | 1 | 251 | |||
| DA498162.1 | - | 1 | 264 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC284898| Gene Symbol: LOC284898 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318329.1 | NP_001305258.1 | 1 | 264 | ||
| GenBank mRNA | AK055980.1 | - | 1 | 44 | ||
| BC036876.1 | - | 1 | 251 | |||
| DA498162.1 | - | 1 | 264 | |||