siRNA Details
| Gene Symbol: TTC28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145418.1 | NP_001138890.1 | 2 | 267 | ||
| XM_005261405.2 | XP_005261462.1 | 2 | 267 | |||
| XM_011530018.2 | XP_011528320.1 | 2 | 446 | |||
| XM_011530019.2 | XP_011528321.1 | 2 | 267 | |||
| XM_011530020.1 | XP_011528322.1 | 2 | 267 | |||
| XM_011530021.2 | XP_011528323.1 | 2 | 267 | |||
| XM_011530022.1 | XP_011528324.1 | 2 | 267 | |||
| XM_017028673.1 | XP_016884162.1 | 2 | 240 | |||
| GenBank mRNA | AB665284.1 | - | 2 | 215 | ||

TTC28
Genomic Map