siRNA Details
| Gene Symbol: FAM150B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002919.2 | NP_001002919.2 | 4 | 695 | ||
| XM_005264689.4 | XP_005264746.1 | 4 | 708 | |||
| XM_006711879.3 | XP_006711942.1 | 4 | 709 | |||
| GenBank mRNA | AY358517.1 | - | 4 | 164 | ||
| BC033490.1 | - | 4 | 569 | |||
| Gene Symbol: LOC101927262 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011510431.2 | XP_011508733.1 | 1 | 56 | ||

FAM150B
Genomic Map