siRNA Details
| Gene Symbol: LOC101929723 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110262.1 | 1 | 307 | |||
| NR_110263.1 | 1 | 307 | ||||
| GenBank mRNA | BX089835.1 | - | 1 | 307 | ||
| CA448932.1 | - | 4 | 603 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929723| Gene Symbol: LOC101929723 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110262.1 | 1 | 307 | |||
| NR_110263.1 | 1 | 307 | ||||
| GenBank mRNA | BX089835.1 | - | 1 | 307 | ||
| CA448932.1 | - | 4 | 603 | |||