siRNA Details
| Gene Symbol: RMND5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022780.3 | NP_073617.1 | 1 | 413 | ||
| GenBank mRNA | AK022815.1 | - | 1 | 390 | ||
| BC012165.1 | - | 1 | 134 | 4636136 | ||
| BC047668.1 | - | 1 | 343 | 6066798 | ||
| Gene Symbol: LOC285074 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026846.1 | 1 | 389 | |||
| GenBank mRNA | BC069048.1 | - | 1 | 171 | ||
| BC132791.1 | - | 1 | 130 | |||
| BC132793.1 | - | 1 | 130 | |||
| BM454994.1 | - | 1 | 342 | |||
| BX353812.2 | - | 1 | 389 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_024288, AK164384, BC050876, CF748545.

RMND5A
Genomic Map