siRNA Details
| Gene Symbol: LOC285095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135766.1 | 2 | 176 | |||
| NR_135767.1 | 2 | 136 | ||||
| NR_135768.1 | 2 | 75 | ||||
| GenBank mRNA | AA250751.1 | - | 2 | 102 | ||
| BX108328.1 | - | 2 | 142 | |||
| DB296165.1 | - | 2 | 448 | |||
| HY129633.1 | - | 2 | 177 | |||

LOC285095
Genomic Map