siRNA Details
| Gene Symbol: LINC00636 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015394.1 | 2, 3 | 275 | |||
| GenBank mRNA | BC101230.2 | - | 2, 3 | 232 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00636| Gene Symbol: LINC00636 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015394.1 | 2, 3 | 275 | |||
| GenBank mRNA | BC101230.2 | - | 2, 3 | 232 | ||