siRNA Details
| Gene Symbol: LINC00692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034055.1 | 6 | 548 | |||
| GenBank mRNA | BC033370.1 | - | 6 | 551 | ||
| BC126324.1 | - | 5 | 385 | |||
| BC143998.1 | - | 5 | 385 | |||
| BC143999.1 | - | 5 | 388 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00692| Gene Symbol: LINC00692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034055.1 | 6 | 548 | |||
| GenBank mRNA | BC033370.1 | - | 6 | 551 | ||
| BC126324.1 | - | 5 | 385 | |||
| BC143998.1 | - | 5 | 385 | |||
| BC143999.1 | - | 5 | 388 | |||