siRNA Details
Gene Symbol: F11-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033901.2 | 3, 4 | 457 | |||
GenBank mRNA | BC038717.1 | - | 3, 4 | 396 | ||
BC117432.1 | - | 2, 3 | 183 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: F11-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033901.2 | 3, 4 | 457 | |||
GenBank mRNA | BC038717.1 | - | 3, 4 | 396 | ||
BC117432.1 | - | 2, 3 | 183 |